Learn More
Buerkle™ Polyethylene Hose Couplings
Shop All Bürkle Products
Click to view available options
Diameter (Metric):
11 to 14 mm
13 to 16 mm
3 to 5 mm
5 to 7 mm
7 to 10 mm
9 to 12 mm
Unit Size:
Pack of 10
Description
- Manufactured from polyethylene
- Provided 10 per pack
- Available for a range of hose diameters
Specifications
Specifications
| Certifications/Compliance | EU Foodstuffs, RoHS |
| Diameter (Metric) | 3 to 5 mm |
| Autoclavable | Autoclavable |
| Material | Polyethylene |
| For Use With | 3 to 5 mm Hose |
| For Use With (Application) | Pressureless Operation |
| Packaging Quantity | 10/Pk. |
| Shape | Conical |
| Product Type | Hose Coupling |