Learn More
Bel-Art™ Polyethylene Utility Bags
Product Code. 15339202
Change view
Click to view available options
Dimensions (L x W):
16 x 10 in. (406 X 254mm)
16 x 8 in. (406 x 203mm)
20 x 12 in. (508 x 305mm)
Thickness (Metric):
0.038 mm
0.102 mm
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Thickness (Metric) unitSize
15339202 16 x 8 in. (406 x 203mm) 0.038 mm Pack of 100
11843921 20 x 12 in. (508 x 305mm) 0.038 mm Pack of 100
18892561 16 x 10 in. (406 X 254mm) 0.102 mm Pack of 100
15582101 16 x 10 in. (406 X 254mm) 0.038 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15339202 Supplier Bel-Art™ Supplier No. H131760816
Add to basket
Add to basket
Reusable, transparent bags are well-suited for shipping or storage and are especially useful as sample bags
  • FDA approved material; Latex free
  • Different sizes available to best match particular packaging requirements
  • Not autoclavable
Specifications
Autoclavable Not Autoclavable
Closure Type Zip Tie
Color Clear
Disposable Single-use
Material Polyethylene
Sterility Non-sterile
Dimensions (L x W) 16 x 8 in. (406 x 203mm)
Length (English) 16 in.
Width (English) 8 in.
Product Type Sample Bag
Length (Metric) 406 mm
Width (Metric) 203 mm
Thickness (Metric) 0.038 mm