missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Polyethylene Wide Mouth Two Color Safety Label Wash Bottle
Description
Wide-Mouth wash bottles for easy refilling.
- Valve eliminates drips — allows dispensing of wash bottle in an upright or inverted position
- Wide mouth 53mm (2 in.) bottle for easy refilling
- Polypropylene closure; color-coded to aid in easy identification
- Translucent, LDPE solvent-resistant
- Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Capacity (English) | 16.9 oz. |
| Color | Translucent |
| Diameter (Metric) Outer Neck | 53mm |
| Includes | Red Polypropylene Cap |
| Material | Polyethylene |
| Quantity | 3/Pk. |
| Sterility | Non-sterile |
| Closure Color | Red |
| Certifications/Compliance | Compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?