missing translation for 'onlineSavingsMsg'
Learn More

Polyethylenimine (Branched) (Technical Grade), TRC

Product Code. 30417964
Change view
Click to view available options
Quantity:
2.5 g
5 g
25 g
Unit Size:
2.5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30417964 2.5 g 2.5g
30417398 25 g 25g
30428383 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30417964 Supplier Toronto Research Chemicals Supplier No. TRCP6885082.5G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25987-06-8
Synonym Ethylenediamine-ethylenimine copolymer, Aziridine-1,2-diaminoethane copolymer, Ethylenediamine, polymer with ethylenimine
IUPAC Name N'-[2-[2-[2-(2-aminoethylamino)ethyl-[2-[bis(2-aminoethyl)amino]ethyl]amino]ethyl-[2-[2-[bis(2-aminoethyl)amino]ethylamino]ethyl]amino]ethyl]ethane-1,2-diamine
SMILES NCCNCCN(CCNCCN(CCN)CCN)CCN(CCNCCN)CCN(CCN)CCN

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C24H63N13/c25-1-7-31-9-18-36(19-12-33-11-17-34(13-3-27)14-4-28)22-24-37(20-10-32-8-2-26)23-21-35(15-5-29)16-6-30/h31-33H,1-30H2
Formula Weight 533.5329
Chemical Name or Material Polyethylenimine (Branched) (Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.