Learn More
Macherey-Nagel™ Polygram CEL 300-10 UV254 Glass Plates
Brand:  Macherey-Nagel™ 808023
Additional Details : Weight : 0.50000kg
Product Code. 10154761
  • 184.00€ / Pack of 25
Estimated Shipment: 17-05-2024
to see stock.
Specifications
25 Pk.
UV254
20 x 20 cm
Glass Plate
0.1 mm