missing translation for 'onlineSavingsMsg'
Learn More

Polyphosphoric Acid, TRC

Product Code. 30413765
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30413765 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30413765

Brand: Toronto Research Chemicals TRCP6894705G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Polyphosphoric Acid
CAS 8017-16-1
Molecular Formula H n+2 P n O 3n+1
Quantity 5 g
Synonym Super Phos 105, Condensed Phosphoric Acids, PPA
SMILES COC(=O)C1=C(O)c2ccccc2S(=O)(=O)N1C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.