Learn More
Chemsplash Polyprop Dust Suit Coverall, Zip Front Navy
Product Code. 30058399
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
30058399 Medium Pack of 50
30058395 Small Pack of 50
30058342 Large Pack of 50
30058213 X-Large Pack of 50
30058303 2X-Large Pack of 50
30058417 3X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 30058399 Supplier Chemsplash Supplier No. 2523.300.005
Only null left
Add to basket
Add to basket
Polypropylene Dust Suit Coverall is made from a 30gsm non-woven material and comes with a full zip front closure, elasticated wrist and ankle to secure fit and has a hood.
Polypropylene Dust Suit Coverall
  • 30GSM Non-Woven Polypropylene
  • Zip Front
  • Elasticated Wrist/Ankle
  • Hood
  • Suitable for Hygiene Rules / Food Processing
  • Clean Areas / Disposal Operations
  • PFAS Free
Description: The Polypropylene Dust Suit Coverall is made from a 30GSM non-woven material and features a full zip front closure, elasticated wrist and ankle for a secure fit, and a hood. Individually packed for hygiene purposes, it is suitable for Hygiene Rules, Food Processing, Clean Areas, and Disposal Operations.
Specifications
Product Type Hooded Coveralls
Size Medium
Gender Unisex
Material Polypropylene
Color Blue
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel
Closure Type Ties
Seam Type Stitched
Sterility Non-sterile
Resistance Particle
Latex Free Yes
No. of Pockets None