missing translation for 'onlineSavingsMsg'
Learn More

Polypropylene (Amorphous), TRC

Product Code. 30011324
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011324 50 g 50g
30009900 5 g 5g
30012766 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011324 Supplier Toronto Research Chemicals Supplier No. TRCP68937550G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9003-07-0
Molecular Formula n(C3H6)
Molecular Weight (g/mol) 42.08
Synonym Polypropylene, Propylene homopolymer, 1-Propene, homopolymer (9CI, ACI)
SMILES CCC(C)C

Specifications

Quantity 50 g
InChI Formula InChI=1S/C3H6/c1-3-2/h3H,1H2,2H3
Formula Weight 42.047
Chemical Name or Material Homopolymer Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.