Learn More
Masterflex™ Polypropylene Barbed Connector/reducer Kits
Product Code. 12832223
Change view
Click to view available options
Description:
Size: Large
Size: Small
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description unitSize
12832223 Size: Large Each
12810553 Size: Small Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12832223 Supplier Masterflex™ Supplier No. 4151407
Only null left
Add to basket
Add to basket
Cole-Parmer Polypropylene Barbed Connector/reducer Kits, Autoclavable: Autoclavable, For Use With: Male Luer Lock, Packaging Quantity: 1 Pack, Barbed Connector/Reducer Kit
Specifications
Description Size: Large
Autoclavable Autoclavable
Material Polypropylene
For Use With Male Luer Lock
Packaging Quantity 1 Pack
Product Type Barbed Connector/Reducer Kit