Learn More
Gilson™ Polypropylene Bottle
Product Code. 11337820 Shop All Gilson Products
Change view
Click to view available options
Volume (Metric):
4 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11337820 4 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11337820 Supplier Gilson™ Supplier No. F1077411
  • ONLINE EXCLUSIVE PRICE
    215.00€
    209.37€ / Pack of 1
    Save 5.63€
    2% Off
Estimated Shipment: 30-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
  • This 4 L polypropylene bottle is used with the Gilson Safe Aspiration Station.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Bottle
Volume (Metric) 4 L