Learn More
Integra Biosciences™ Polypropylene, Bottle and Lid with Tubing Fittings
Product Code. 10163703
Change view
Click to view available options
Capacity (Metric):
4 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10163703 4 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10163703 Supplier Integra Biosciences™ Supplier No. 158432
  • ONLINE EXCLUSIVE PRICE
    197.00€
    182.03€ / Pack of 1
    Save 14.97€
    7% Off
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Capacity Metric: 4 L
  • 4L Polypropylene bottle and lid
  • With tubing fittings
  • For use with VACUSAFE Aspiration Systems
VACUSAFE Aspiration System is a compact, all-in-one laboratory vacuum pump, providing a safe and efficient way to collect and contain biological liquid waste.
Specifications
Capacity (Metric) 4 L
Coating Polypropylene
Material Polypropylene
Product Type Reservoir Bottle