Learn More
Chemsplash Polypropylene Boxer Shorts
Product Code. 30058212
Change view
Click to view available options
Size:
Standard
Unit Size:
Pack of 250
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
30058212 Standard Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30058212 Supplier Chemsplash Supplier No. 2784.400.999
  • ONLINE EXCLUSIVE PRICE
    139.52€
    129.09€ / Pack of 250
    Save 10.43€
    7% Off
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This product is suitable for use in BRC approved work areas and for the Pharmaceutical Industry.
0 GSM Polypropylene Boxer Shorts
  • Seams Overlocked
  • Elasticated Waist
  • Standard Size: 51x37 cm
  • Size XL: 60x44 cm
  • 10pcs/polybag, 250pcs/ctn
  • PFAS Free
Description: These 0 GSM Polypropylene Boxer Shorts are designed for use in BRC approved work areas and the Pharmaceutical Industry. The boxer shorts feature overlocked seams and an elasticated waist, providing a comfortable fit. They come in two sizes: Standard (51x37 cm) and XL (60x44 cm). The product is PFAS free, ensuring it meets industry standards for safety and hygiene. The boxer shorts are packed in polybags, with 10 pieces per bag and 250 pieces per carton.
Specifications
Size Standard
Color White