Learn More
VITLAB™ Polypropylene Burette Clamps
Brand:  VITLAB™ 80139
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15272719
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Specifications
Polypropylene
Rubber-Coated Tips and Springs
Buret Clamp
8 to 14 mm