Learn More
Cole-Parmer™ Polypropylene Magnetic Drive Centrifugal Pump
Product Code. 15214058
Change view
Click to view available options
Height (Metric):
105 mm
130 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) unitSize
15214058 105 mm Each
15204257 130 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15214058 Supplier Cole-Parmer™ Supplier No. 7201045
Only null left
Specifications
Type Magnetic Drive Centrifugal Pump
Material Polypropylene
Length (Metric) 257 mm
Height (Metric) 105 mm
Width (Metric) 121 mm
Certifications/Compliance CE Compliance
Voltage 220 VAC