Learn More
Vitlab™ Polypropylene Measuring Scoops
Vitlab™ Polypropylene Measuring Scoops with precisely formed filling edge and comfortable, stable handle.
Brand:  Vitlab™ 396952
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 15372974
  • 19.80€ / Pack of 12
Estimated Shipment: 14-06-2024
to see stock.
Add to basket
Description
Description
  • Measuring scoops in different colours
  • Also suitable as weighing scoops
  • With precisely formed filling edge and comfortable, stable handle
  • Easily readable volume specification on the upper side of the handle
Specifications
Specifications
Polypropylene
Green
200 mm
100 mL
Foodsafe according to EU Directive No. 10/2011