Learn More
Vitlab™ Polypropylene Measuring Scoops
Vitlab™ Polypropylene Measuring Scoops with precisely formed filling edge and comfortable, stable handle.
Brand: Vitlab™ 396940
Additional Details : Weight : 0.50000kg
Description
- Measuring scoops in different colours
- Also suitable as weighing scoops
- With precisely formed filling edge and comfortable, stable handle
- Easily readable volume specification on the upper side of the handle
Specifications
Polypropylene | |
Red | |
200 mm |
100 mL | |
Foodsafe according to EU Directive No. 10/2011 |