Learn More
Greiner Bio-One Polypropylene Micro Pipet Tips
Designed for precise pipetting of the smallest possible liquid volumes. Greiner Bio-One™ Polypropylene Micro Pipet Tips provide a small dead volume due to their small size.
Brand:  Greiner Bio-One 771287
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 10115201
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • Manufactured from high-grade polypropylene for optimum performance and fit
  • High breaking strength, dimensionally stable
  • Heat resistant to +140°C 
  • Free of heavy metals
  • Available in bulk pacaging or convenient rack
  • Conform to ISO 9001 standard
Specifications
Specifications
0.5 to 10 μL
Natural
P10 standard
Gilson™ Pipetman™ U10, Gilson™ Pipetman™ P10, Gilson™ Pipetman™ P2
Non-sterile
Autoclavable
4800/Pack