Learn More
Greiner Bio-One Polypropylene Micro Pipet Tips
Designed for precise pipetting of the smallest possible liquid volumes. Greiner Bio-One™ Polypropylene Micro Pipet Tips provide a small dead volume due to their small size.
Brand: Greiner Bio-One 771287
Additional Details : Weight : 0.80000kg
Description
- Manufactured from high-grade polypropylene for optimum performance and fit
- High breaking strength, dimensionally stable
- Heat resistant to +140°C
- Free of heavy metals
- Available in bulk pacaging or convenient rack
- Conform to ISO 9001 standard
Specifications
0.5 to 10 μL | |
Natural | |
P10 standard | |
Gilson™ Pipetman™ U10, Gilson™ Pipetman™ P10, Gilson™ Pipetman™ P2 |
Non-sterile | |
Autoclavable | |
4800/Pack | |
Polypropylene |