Learn More
Fisherbrand™ Polypropylene Microtubes with Snap Cap
Provide a safe, liquid-tight, reliable seal even with prolonged boiling using Fisherbrand™ Polypropylene Microtubes. Use these polypropylene tubes for sample preparation, centrifugation or storage.The microtubes provide convenient, ergonomic and reliable one-handed operation.
Brand: Fisherbrand™
Additional Details : Weight : 0.95000kg
Description
- Flat snap cap
- Autoclavable
- Polypropylene
- Eppendorf style
Specifications
10 mm | |
Polypropylene | |
1.5 mL | |
Polypropylene | |
1000/Pk. | |
No |
Flat cap | |
Yes | |
Red | |
40mm | |
1.5mL |