Description
- Polypropylene/polycarbonate construction
- Holds up to 7 Gilson single or multichannel pipettors
- Supplied as pack of 8 stands
Specifications
Specifications
| Material | Polycarbonate/Polypropylene |
| Product Line | CARROUSEL |
| Type | Pipette Stand |
| For Use With (Equipment) | Gilson Pipette |
| Quantity | 8 Pk. |