missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ Polypropylene Reagent Bottle

Product Code. 12895733
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
2000 mL
Closure Type:
Screw Cap with GL Thread
Stopper with Hexagonal Cap
Thread Size (Metric):
GL 25
GL 32
GL 45
GL 63
NS
Unit Size:
Pack of 10
Pack of 20
Pack of 6
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Type Thread Size (Metric) Unit Size
12895733 1000 mL Screw Cap with GL Thread GL 45 Pack of 10
15712046 100 mL Stopper with Hexagonal Cap NS Pack of 20
15291169 100 mL Screw Cap with GL Thread GL 32 Pack of 20
15722046 250 mL Stopper with Hexagonal Cap NS Pack of 20
15271159 500 mL Screw Cap with GL Thread GL 25 Pack of 10
10443101 500 mL Stopper with Hexagonal Cap NS Pack of 10
15281159 1000 mL Screw Cap with GL Thread GL 32 Pack of 10
15732046 1000 mL Stopper with Hexagonal Cap NS Pack of 10
15211179 2000 mL Screw Cap with GL Thread GL 63 Pack of 6
10641353 2000 mL Screw Cap with GL Thread GL 32 Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 12895733 Supplier VITLAB™ Supplier No. 102089
  • ONLINE EXCLUSIVE PRICE
    80.90€
    69.20€ / Pack of 10
    Save 11.70€
    14% Off
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Store liquid and powder chemicals securely. VITLAB™ Polypropylene Reagent Bottle is constructed of transparent, durable polypropylene for long-term storage. Choose from neck-size stoppers or maximum grip screw caps, both available in a variety of sizes.

Economical and durable plastic reagent bottles are ideal of storing liquids, powders and paste-like materials. A variety of sizes, seals (stopper or screw caps) and neck sizes are available to meet the storage needs of any lab material.
  • Transparent media bottles ideal for long-term storage of chemicals
  • Reagent bottles featuring screw caps or neck stoppers
    • Stoppers: neck-size with a hexagonal-knob cap and red core
    • Screw caps: high-pitch GL threads for significant carrying power
  • Larger sizes (5 and 10L) available with handles for easier carrying
  • Wider mouth bottles are ideal for easier filling, especially of powders and paste-like materials
  • Constructed of study polypropylene
    • Good chemical and temperature resistance
    • Autoclavable at 121 °C (2 bar) according to DIN EN 285
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1000 mL
For Use With (Application) Long-term storage of liquids
Includes Screw cap
Material Polypropylene
No. per Pack 10 Bottles
Thread Size (Metric) GL 45
Mouth Wide
Height (Metric) 197 mm
Diameter (Metric) 105 mm
Closure Type Screw Cap with GL Thread
Autoclavable Autoclavable
Product Type Reagent Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.