Learn More
BRAND™ Polypropylene Reservoir
Product Code. 15922957 Shop All Brand Products
Change view
Click to view available options
Capacity (Metric):
220 mL/Column
40 mL/Column
50 mL/Column
6 mL/Column
60 mL/Column
Channels:
1
4
6
12
Unit Size:
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Channels unitSize
15922957 220 mL/Column 1 Pack of 10
15932957 6 mL/Column 12 Pack of 10
15942957 60 mL/Column 4 Pack of 10
15952957 40 mL/Column 6 Pack of 10
15962957 50 mL/Column 1 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15922957 Supplier BRAND™ Supplier No. 701450
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Reservoirs allow for different volumes and different solutions to be used in the automated liquid handling process.
  • Pyramidal or flat bottom
  • DNase-free
  • RNase-free
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance DIN EN 285
Description 96-well
Capacity (Metric) 220 mL/Column
Material Polypropylene
Sterility Non-sterile
Volume (Metric) Well 220 mL
Autoclavable Autoclavable
Channels 1
Bottom Shape Conical