missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Polypropylene Screw Caps

Product Code. 11683129 Shop All Brand Products
Change view
Click to view available options
Product Type:
Screw Cap
Tamper-evident Screw Cap
Thread Size (Metric):
GL 25
GL 32
GL 45
GL 54
GL 80
Unit Size:
Each
Pack of 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Thread Size (Metric) Unit Size
11683129 Tamper-evident Screw Cap GL 54 Each
15281249 Screw Cap GL 25 Pack of 10
15291249 Screw Cap GL 32 Pack of 10
11603129 Tamper-evident Screw Cap GL 32 Each
15201259 Screw Cap GL 45 Pack of 10
11693129 Tamper-evident Screw Cap GL 80 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11683129 Supplier BRAND™ Supplier No. 130324
  • ONLINE EXCLUSIVE PRICE
    1.18€
    0.92€ / Pack of 1
    Save 0.26€
    22% Off
View all promotions
In Stock
Add to basket
Add to basket

Manufactured from polypropylene. BRAND™ Polypropylene Screw Caps are available in a variety of sizes. Pouring rings sold separately.

  • Manufactured from polypropylene.
  • Replacement and spare screw caps for laboratory bottles available in a variety of sizes.
  • Pouring rings sold separately.
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Material Polypropylene
Quantity 100/Pk.
Thread Size (Metric) GL 54
Certified Yes
Neck Finish 30 mm
Mouth Wide
For Use With (Equipment) For use with wide-neck bottles
Product Type Tamper-evident Screw Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.