Learn More
Multiroir™ Polypropylene Storage Boxes
Store and transport items securely in high-capacity Multiroir™ Polypropylene Storage Boxes. The boxes have snap-on, tight-fitting lids and have no handles.
Brand: Multiroir™ 18477
Additional Details : Weight : 0.80000kg
Description
- Translucent Polypropylene
- Large capacity
- Without handles
Specifications
30 L | |
Polypropylene | |
Storage Box | |
413 x 345 x 294 mm | |
345 mm | |
294 mm |
Translucent | |
Rectangle | |
1 Each | |
413 mm | |
345 mm | |
1.17 kg |