Learn More
Bel-Art™ SP Scienceware™ Polypropylene Thermometer Joint Adapter
Product Code. 10062554
Change view
Click to view available options
For Use With (Application):
For 14/20 Tapered Joints
For 24/40 Tapered Joints
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
10062554 For 24/40 Tapered Joints Each
10053274 For 14/20 Tapered Joints Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10062554 Supplier Bel-Art™ SP Scienceware™ Supplier No. F203091716
  • ONLINE EXCLUSIVE PRICE
    127.00€
    102.57€ / Pack of 1
    Save 24.43€
    19% Off
Estimated Shipment: 15-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Joint Adapters have a Teflon™ PTFE body with Viton™ O-ring for air tight vacuum work
  • Adapter has a threaded polypropylene screw cap on top with an inside O-ring for depth adjustment of 6-7mm for straight stem thermometers, glass tubing and other items
  • Rotating the extracting nut removes the adapter from flask
  • Not autoclavable
Specifications
Depth (Metric) 6 to 7 mm
For Use With (Application) For 24/40 Tapered Joints
Includes Screw cap with inside O-ring
Material Polypropylene, PTFE
Product Type Thermometer Joint Adapter
For Use With (Equipment) Straight Stem Thermometers, Glass Tubing