Learn More
Haleco™ Polypropylene Universal Absorbent Sheets
Product Code. 15437640
Change view
Click to view available options
Material:
Polypropylene
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
15437640 Polypropylene Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15437640 Supplier Haleco™ Supplier No. UMU114H
  • ONLINE EXCLUSIVE PRICE
    66.50€
    60.12€ / Pack of 1
    Save 6.38€
    9% Off
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Color Gray
Material Polypropylene
Packaging Quantity 1 Ea.
Length (Metric) 2300 cm
Height (Metric) 38 cm
Weight (Metric) 2.8 kg