missing translation for 'onlineSavingsMsg'
Learn More

Polyquaternium 1, TRC

Product Code. 30005210
Change view
Click to view available options
Quantity:
100 mg
500 mg
5 g
Unit Size:
100mg
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005210 5 g 5g
30011574 500 mg 500mg
30006851 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005210 Supplier Toronto Research Chemicals Supplier No. TRCP6881005G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75345-27-6
Molecular Formula (C6H12ClN)n.C16H36Cl2N2O6
Molecular Weight (g/mol) 556.99
Synonym α-[4-[Tris(2-hydroxyethyl)ammonio]-2-buten-1-yl]-ω-[tris(2-hydroxyethyl)ammonio]poly[(dimethyliminio)-2-butene-1,4-diyl Chloride] Chloride, Onamer M, Onyxsperse 12S, Polidronium Chloride, Polyquad, Polyquat 1;
IUPAC Name dimethyl-bis[(E)-4-[tris(2-hydroxyethyl)azaniumyl]but-2-enyl]azanium;trichloride
SMILES C[N+](C/C=C/C[N+](CCO)(CCO)CCO)(C/C=C/C[N+](CCO)(CCO)CCO)C.[Cl-].[Cl-].[Cl-]

Specifications

Quantity 5 g
InChI Formula InChi=1S/C22H48N3O6.3ClH/c1-23(2,7-3-5-9-24(11-17-26,12-18-27)13-19-28)8-4-6-10-25(14-20-29,15-21-30)16-22-31;;;/h3-6,26-31H,7-22H2,1-2H3;3*1H/q+3;;;/p-3/b5-3+,6-4+;;;
Formula Weight 555.2609
Chemical Name or Material Polyquaternium 1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.