Learn More
Greiner Bio-One Polystyrene Conical Bottom Analyzer Cups
Product Code. 15975311
Change view
Click to view available options
Volume (Metric):
4 mL
For Use With (Equipment):
Technicon Analyzer
Unit Size:
Pack of 1000
Pack of 5000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) For Use With (Equipment) Unit Size
15975311 4 mL Technicon Analyzer Pack of 5000
10660742 4 mL Technicon Analyzer Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15975311 Supplier Greiner Bio-One Supplier No. 621171
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Greiner Bio-One Cell 1.5mL Polystyrene Conical Bottom Analyzer Cups suitable for use with analytical systems, such as Technicon, Gemsaec, Centrifichem, Hitachi, TOA and Coulter analyzers.
  • Material: Polystyrene
  • Conical bottom
  • With support skirt
  • For single use only
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Technicon Analyzer
Shape Conical
Sterility Non-sterile
Color Clear
Material Polystyrene
Product Type Analyzer Cup
Volume (Metric) 4 mL