Learn More
mb2 Polyurethane Chair on Castors
Product Code. 11927834
Change view
Click to view available options
Dimensions: 610 dia. x 480mmH
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11927834 Dimensions: 610 dia. x 480mmH
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11927834 Supplier mb2 Supplier No. AP1B2D1NE2
  • ONLINE EXCLUSIVE PRICE
    282.00€
    251.00€ / Pack of 1
    Save 31.00€
    10% Off
In Stock
Add to basket
Add to basket
Polyurethane chair on castors - Mid lift - Fix mechanism
  • Chair in polyurethane
  • Seat and back in polyurethane
  • Backrest adjustable in depth by wheel.
  • Pneumatic cylinder mid
  • 5-spoke base injected in nylon and fiberglass reinforced by a galvanized steel ring.
  • Hard floor castors 50 mm
Specifications
Product Type Contoured Seat
Color Black
Adjustable (Height) Yes
Adjustable Height
Autoclavable Not Autoclavable
Back Style Standard
Base Style Branched 5 Wheel
Diameter (Metric) 610 mm
For Use With (Application) For swivel chair
Height (Metric) 700 mm
Quantity 1 Ea.
Seating Form Contoured
Seating Height 500 to 690 mm
Weight (Metric) 10.50 kg
With Armrest No
With Backrest Yes
With Casters Yes
Footrest No
With Lumbar Support No
With Tilt No
Material Polyurethane
Seating Type Stool with Back