missing translation for 'onlineSavingsMsg'
Learn More

Pongamol, TRC

Product Code. 30132728
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
Unit Size:
10mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132728 250 mg 250mg
30137243 50 mg 50mg
30011175 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132728 Supplier Toronto Research Chemicals Supplier No. TRCP699078250MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 484-33-3
Molecular Formula C18 H14 O4
Molecular Weight (g/mol) 294.3
Synonym 1-(4-methoxy-5-benzofuranyl)-3-phenyl-1,3-propanedione, Lanceolatin C
IUPAC Name 1-(4-methoxybenzofuran-5-yl)-3-phenyl-propane-1,3-dione
SMILES COc1c(ccc2occc12)C(=O)CC(=O)c3ccccc3

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C18H14O4/c1-21-18-13(7-8-17-14(18)9-10-22-17)16(20)11-15(19)12-5-3-2-4-6-12/h2-10H,11H2,1H3
Formula Weight 294.0829
Chemical Name or Material Pongamol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.