Learn More
Heinz Herenz Porcelain Pestle with Rough Finish
Product Code. 30158503
Change view
Click to view available options
Capacity (Metric):
54 mm
115 mm
135 mm
150 mm
175 mm
210 mm
250 mm
280 mm
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
30158503 175 mm Each
30158514 210 mm Each
30158433 150 mm Each
30158356 115 mm Each
30158457 - Each
30158430 280 mm Each
30158466 135 mm Each
30158479 250 mm Each
30158321 54 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 30158503 Supplier Heinz Herenz Supplier No. 1070978
Add to basket
Add to basket
Porcelain pestle with rough finish.
  • Rough finish
  • Porcelain
Specifications
Autoclavable Autoclavable
Product Type Pestle
Material Porcelain
Type Pestle
Finish Rough
Capacity (Metric) 175 mm