missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Porimin Antibody (CL14381) [mFluor Violet 610 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP3-26852MFV610
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Porimin Monoclonal antibody specifically detects Porimin in Human samples. It is validated for Immunohistochemistry
Especificaciones
| Porimin | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| KCT-3, KCT3keratinocytes associated transmembrane protein 3, Keratinocytes-associated transmembrane protein 3, PORIMINporimin, pro oncosis receptor inducing membrane injury, Pro-oncosis receptor inducing membrane injury, serine/threonine-rich receptor, transmembrane protein 123PORMIN | |
| This antibody was developed using a synthetic peptide derived from Q8N131, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Apoptosis | |
| 114908 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Immunohistochemistry | |
| CL14381 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido