Learn More
Hanna Instruments™ Portable EC/TDS/Temperature Meter
Product Code. 11707964
Change view
Click to view available options
Range (Conductivity):
0 to 3999 μS/cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range (Conductivity) Unit Size
11707964 0 to 3999 μS/cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 11707964 Supplier Hanna Instruments™ Supplier No. HI99300N
  • ONLINE EXCLUSIVE PRICE
    674.15€
    270.49€ / Pack of 1
    Save 403.66€
    59% Off
View all promotions
Estimated Shipment: 08-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Waterproof case and large, dual level LCD screen
  • Portable, waterproof Portable Low Range EC/TDS Meter
  • Suitable for use in applications as diverse as wastewater treatment, reverse osmosis, and HVAC
  • Measures low range conductivity in μS/cm and TDS in ppm
  • Large multi-level LCD displays the primary reading, temperature, and calibration guides simultaneously
  • One button automatic calibration (conductivity or TDS)
  • Automatic temperature compensation with adjustable temperature correction coefficient, β
Specifications
Accuracy (Conductivity) ±2% Full-scale
Calibration Automatic, one point at 1413 μS/cm, 1382 ppm (CONV 0.5) or 1500 ppm (CONV 0.7)
Calibration Points 1
Range (Conductivity) 0 to 3999 μS/cm
Resolution (Conductivity) 1 μS/cm
Temperature Compensation Automatic, 0 to 60°C (32 to 140°F) with β adjustable from 0.0 to 2.4%/°C with 0.1%
Range (TDS) 0 to 2000 ppm (mg/L)
Resolution (TDS) 1 ppm (mg/L)
Accuracy (TDS) ±2% Full-scale
Resolution (Temperature) 0.1°C (0.1°F)
Accuracy (Temperature) ±0.5°C
Display Type LCD
Dimensions (L x W x H) 152 x 58 x 30 mm
Weight (Metric) 205 g
Warranty 2 Years
Probe Type HI 76306 EC/TDS probe with internal temperature sensor, DIN connector and 1 m (3.3’) cable (included)
Type Portable EC/TDS/Temperature Meter
Battery Life Approximately 500 hr. of continuous use, auto-off after eight min. of non-use
Voltage 1.5 V