Learn More
Hughes Portable Emergency Eye/Face/Body Wash
Product Code. 17752338
Change view
Click to view available options
Height (Metric):
24 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
17752338 24 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17752338 Supplier Hughes Supplier No. HEWS0038GV
  • ONLINE EXCLUSIVE PRICE
    1476.09€
    928.07€ / Pack of 1
    Save 548.02€
    37% Off
View all promotions
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Portable, stainless steel emergency eye, face and body shower.
  • This portable 15 liter stainless steel cylinder with Optiflex hand shower effectively washes eyes, face and body delivering up to three minutes of water when pressurised via a Schraeder valve
  • The pressure gauge and dual action safety valve prevent over pressurisation and allow the user to vent the cylinder for cleaning and maintenance
Specifications
Height (Metric) 24 cm
Material Stainless Steel
Product Type Emergency Wash
Width (Metric) 26.7 cm