Learn More
Esculape Laboratoire™ Eyewash Bottles for First Aid
Product Code. 11749522
Change view
Click to view available options
Volume (Metric):
200 mL
500 mL
Composition:
Sterile Isotonic Solution
Sterile Ophthalmic Solution
Includes:
200 mL Blue Neutral pH Solution, 50 0mL Bottle Green Sterile Ophthalmic Solution
Cap
Cap, Storage Case, Belt Loop w/ Carrying Case
Cap, Storage, Carrying Case
Quantity:
10 Pk.
12 Pk.
Ea.
Unit Size:
Each
Pack of 10
Pack of 12
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Composition Includes Quantity Unit Size
11749522 500 mL Sterile Ophthalmic Solution Cap 12 Pk. Pack of 12
11769102 200 mL Sterile Isotonic Solution Cap, Storage, Carrying Case Ea. Each
11962875 200 mL Sterile Isotonic Solution Cap, Storage Case, Belt Loop w/ Carrying Case Ea. Each
11759522 200 mL Sterile Ophthalmic Solution 200 mL Blue Neutral pH Solution, 50 0mL Bottle Green Sterile Ophthalmic Solution Ea. Each
11997625 200 mL Sterile Ophthalmic Solution Cap Ea. Each
11891553 200 mL Sterile Ophthalmic Solution Cap 10 Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11749522 Supplier Esculape Laboratoire™ Supplier No. 8205259
  • ONLINE EXCLUSIVE PRICE
    418.95€
    178.21€ / Pack of 12
    Save 240.74€
    57% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Use Esculape™ Eyewash Bottles for First Aid has a carrying case with a belt loop for portabilitiy.
  • Storage case
  • Contain sterile isotonic solution
  • Ergonomic
Specifications
Product Type Eyewash Bottle
Composition Sterile Ophthalmic Solution
For Use With (Application) Portable Eyewash
Includes Cap
Quantity 12 Pk.
Sterility Sterile
Volume (Metric) 500 mL