Learn More
Gilson™ Position Rack
Product Code. 16666125 Shop All Gilson Products
Change view
Click to view available options
For Use With (Equipment):
5 mL Vial (12 x 75/100 mm)
9 mL Vial (13 x 100 mm)
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
16666125 9 mL Vial (13 x 100 mm) Each
16686125 5 mL Vial (12 x 75/100 mm) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16666125 Supplier Gilson™ Supplier No. 170413
 Restricted Item
In Stock
Add to basket
Add to basket
These racks are for use with the FC203B and FC204 Fraction Collectors.
  • HPLC Purification Technique
  • Fraction Collector Racks
Specifications
Description Code 11
For Use With (Equipment) 9 mL Vial (13 x 100 mm)
No. of Positions 105