Learn More
Brady™ Posts and Fillable Bases
Product Code. 11701306
Change view
Click to view available options
Color:
Black and Yellow Striped
Red
Red on White
White
Unit Size:
1 pack
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11701306 Red on White 1 pack
11711306 Black and Yellow Striped 1 pack
11741306 White 1 pack
11761306 Red 1 pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11701306 Supplier Brady™ Supplier No. 256721
  • ONLINE EXCLUSIVE PRICE
    119.00€
    106.84€ / Pack of 1
    Save 12.16€
    10% Off
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
These Posts and Fillable Bases are ideal for controlling and directing foot traffic. Suitable for warehouses, factories, offices etc. Chains and ribbons can be attached to the sturdy 900 mm posts.
  • Ideal for controlling and directing foot traffic.
  • Suitable for warehouses, factories, offices etc.
  • Chains and ribbons can be attached to the sturdy 900 mm posts.
  • Posts and Fillable Bases.
Specifications
Alternate Part Number POST & FILLABLE BASE RED/WHT PK 4
Diameter (Metric) Outer 40 mm
Height (English) 35.43 in.
Height (Metric) 900 mm
Packaging Quantity 4 Pk.
Color Red on White
Type Post with Fillable Base