Learn More
Potassium 2,4-dichlorophenyltrifluoroborate, 96%, Thermo Scientific™
Product Code. 15449857
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15449857 1 g 1g
15459857 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15449857 Supplier Thermo Scientific Alfa Aesar Supplier No. H53083.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 192863-38-0
Molecular Formula C6H3BCl2F3K
Molecular Weight (g/mol) 252.89
MDL Number MFCD04115745
InChI Key MASOPVJEJIORDH-UHFFFAOYSA-N
Synonym potassium 2,4-dichlorophenyl trifluoroborate, potassium 2,4-dichlorophenyl trifluoroboranuide, potassium 2,4-dichlorophenyltrifluoroborate, potassiotrifluoro 2,4-dichlorophenyl boron v, potassium 3-dichlorophenyl trifluoroborate, potassium 2,4-dichlorophenyl-trifluoroboranuide, potassium ion 2,4-dichlorophenyl trifluoroboranuide, potassium 2,4-dichlorophenyl trifluoro borate 1-, potassium 2,4-dichlorophenyl-tris fluoranyl boranuide
PubChem CID 23672565
IUPAC Name potassium;(2,4-dichlorophenyl)-trifluoroboranuide
SMILES [K+].F[B-](F)(F)C1=C(Cl)C=C(Cl)C=C1
Specifications
Melting Point 98°C to 104°C
Quantity 1 g
Formula Weight 252.9
Percent Purity 96%
Chemical Name or Material Potassium 2,4-dichlorophenyltrifluoroborate