missing translation for 'onlineSavingsMsg'
Learn More

Potassium 2-methylphenyltrifluoroborate, 96%, Thermo Scientific™

Product Code. 15479967
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15479967 1 g 1g
15489967 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15479967 Supplier Thermo Scientific Alfa Aesar Supplier No. H53214.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 274257-34-0
Molecular Formula C7H7BF3K
Molecular Weight (g/mol) 198.04
MDL Number MFCD03790006
InChI Key ROMUTKWSUQKVLS-UHFFFAOYSA-N
Synonym potassium trifluoro o-tolyl borate, potassium 2-methylphenyl trifluoroborate, potassium o-tolyltrifluoroborate, potassium trifluoro 2-methylphenyl boranuide, potassium 2-methylphenyltrifluoroborate, pubchem11432, potassiumo-tolyltrifluoroborate, trifluoro 2-methylphenyl potassioboron v, potassium trifluoro-2-methylphenyl boranuide, potassium trifluoro 2-methylphenyl borate 1-
PubChem CID 23674703
IUPAC Name potassium;trifluoro-(2-methylphenyl)boranuide
SMILES [K+].CC1=CC=CC=C1[B-](F)(F)F

Specifications

Melting Point 90°C to 94°C
Quantity 1 g
Formula Weight 198.04
Percent Purity 96%
Chemical Name or Material Potassium 2-methylphenyltrifluoroborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.