missing translation for 'onlineSavingsMsg'
Learn More

Potassium (3-chlorophenyl)trifluoroborate, 97%, BLD Pharm™

Product Code. 30988045
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
Unit Size:
25g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30988045 5 g -
30983419 250 mg -
30983141 25 g 25g
30973870 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30988045 Supplier BLD Pharm Supplier No. BD1486625g
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 411206-75-2
Molecular Formula C6H4BClF3K
MDL Number MFCD04115754
Quantity 5 g
SMILES F[B-](F)(C1=CC=CC(Cl)=C1)F.[K+]
Molecular Weight (g/mol) 218.45
Percent Purity 97%
Chemical Name or Material Potassium (3-chlorophenyl)trifluoroborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.