missing translation for 'onlineSavingsMsg'
Learn More

Potassium 4-methylphenyltrifluoroborate, 98%

Product Code. 11378546
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378546 1 g 1g
11388546 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378546 Supplier Thermo Scientific Alfa Aesar Supplier No. L17604.03
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 216434-82-1 | C7H7BF3K | 198.037 g/mol

In Suzuki Cross-Coupling organotrifluoroborates is used as potent boronic acid surrogates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In Suzuki Cross-Coupling organotrifluoroborates is used as potent boronic acid surrogates.

Solubility
Soluble in water.

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 216434-82-1
Molecular Formula C7H7BF3K
Molecular Weight (g/mol) 198.037
MDL Number MFCD01318171
InChI Key KRWDYXJWQBTBAH-UHFFFAOYSA-N
Synonym potassium p-tolyltrifluoroborate, potassium trifluoro p-tolyl borate, potassium 4-methylphenyl trifluoroborate, potassium 4-methylphenyltrifluoroborate, potassium trifluoro 4-methylphenyl boranuide, 4-methylphenyltrifluoroboric acid potassium salt, pubchem11452, trifluoro 4-methylphenyl potassioboron v, potassium trifluoro p-tolyl boranuide, potassium trifluoro-4-methylphenyl boranuide
PubChem CID 23682258
IUPAC Name potassium;trifluoro-(4-methylphenyl)boranuide
SMILES [B-](C1=CC=C(C=C1)C)(F)(F)F.[K+]

Specifications

Melting Point >300°C
Quantity 1 g
Beilstein 8290098
Solubility Information Soluble in water.
Formula Weight 198.04
Percent Purity 98%
Chemical Name or Material Potassium 4-methylphenyltrifluoroborate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.