missing translation for 'onlineSavingsMsg'
Learn More

Potassium acetate, 8M aq. soln., RNAse free

Product Code. 15445159
Change view
Click to view available options
Quantity:
125 mL
250 mL
Unit Size:
125mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15445159 125 mL 125mL
15455159 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15445159 Supplier Thermo Scientific Alfa Aesar Supplier No. J62856.=E

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 127-08-2 | C2H3KO2 | 98.142 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Potassium Acetate is a potassium salt employed to replenish electrolytes, for restoration of water-electrolyte balance, as well as a urinary and systemic alkalizer. Potassium Acetate can be administered orally or by intravenous infusion. Formerly, It was employed in diuretics and expectorants. A potassium source employed in DNA and protein purification. Some other applications includes as buffering agent, neutralizer, preservative, color retention agent(protect natural color of animals and plants).

Solubility
Soluble in water (98.2 g/L completely soluble) at 25°C, alcohol, methanol, and ammonia. Insoluble in acetone, and ether.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-08-2
Molecular Formula C2H3KO2
MDL Number MFCD00012458
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
Synonym potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
PubChem CID 517044
ChEBI CHEBI:32029
IUPAC Name potassium;acetate

Specifications

Color Colorless
Physical Form Liquid
Quantity 125 mL
Linear Formula CH3CO2K
SMILES CC(=O)[O-].[K+]
Molecular Weight (g/mol) 98.142
Formula Weight 98.15
Concentration or Composition (by Analyte or Components) 8M aq. soln.
Chemical Name or Material Potassium acetate, RNAse free

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.