missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromate solution, 0.0083M EU Pharmacopoeia, Fisher Chemical™

Product Code. 12920874 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12920874 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12920874 Supplier Fisher Chemical Supplier No. VL 034 1000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

Packaging Bottle
CAS 1-2-7758
Molecular Formula BrKO3
Concentration 0.0083 M
InChI Key OCATYIAKPYKMPG-UHFFFAOYSA-M
SMILES [O-]Br(=O)=O.[K+]
IUPAC Name potassium;bromate
Molecular Weight (g/mol) 167.00
PubChem CID 23673461
Quantity 1000 mL
ChEBI CHEBI:38211
Grade Eu. Ph. Reference
Chemical Name or Material Potassium bromate solution
Show More Show Less
compliance-icons
Product Identifier
  • Potassium bromate 0.0083M. Volumetric Solution European Pharmacopoeia Ref: 3004500
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.