missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromide, TRC

Product Code. 30003519
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003519 10 g 10g
30010694 100 g 100g
30004697 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003519 Supplier Toronto Research Chemicals Supplier No. TRCP69504510G
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-02-3
Molecular Formula Br K
Molecular Weight (g/mol) 119
Synonym Potassium bromide, NSC 77367, Potassium bromide, NSC 77367
IUPAC Name potassium;bromide
SMILES [K+].[Br-]

Specifications

Quantity 10 g
InChI Formula InChI=1S/BrH.K/h1H;/q;+1/p-1
Formula Weight 117.882
Chemical Name or Material Potassium bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.