missing translation for 'onlineSavingsMsg'
Learn More

Potassium Citrate Monobasic, TRC

Product Code. 30418462
Change view
Click to view available options
Quantity:
50 g
250 g
500 g
Unit Size:
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30418462 500 g -
30426090 250 g 250g
30412929 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30418462 Supplier Toronto Research Chemicals Supplier No. TRCP698710500G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 866-83-1
Molecular Formula C6H7KO7
Molecular Weight (g/mol) 230.21
Synonym Monobasic Potassium Citrate, Monopotassium Citrate, Potassium Citrate, Potassium Dihydrogen Citrate, 2-Hydroxy-1,2,3-propanetricarboxylic Acid Potassium Salt, 1,2,3-Propanetricarboxylic acid, 2-hydroxy-, potassium salt (1:1), 1,2,3-Propanetricarboxylic acid, 2-hydroxy-, monopotassium salt (9CI), Citric acid, monopotassium salt (8CI), Potassium citrate (6CI), Potassium citrate (KH2(O7C6H5)) (7CI)
IUPAC Name potassium;3-carboxy-3,5-dihydroxy-5-oxopentanoate
SMILES [K+].OC(=O)CC(O)(CC(=O)[O-])C(=O)O

Specifications

Quantity 500 g
InChI Formula InChI=1S/C6H8O7.K/c7-3(8)1-6(13,5(11)12)2-4(9)10;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);/q;+1/p-1
Formula Weight 229.9829
Chemical Name or Material Potassium Citrate Monobasic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.