missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Dodecanoate-d23, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Potassium Dodecanoate-D23 |
| CAS | 51732-20-8 |
| Molecular Formula | C12 D23 O2 . K |
| Quantity | 0.25 g |
| Synonym | Dodecanoic-d23 acid, potassium salt (9 CI), Perdeuterododecanoic acid potassium salt, Potassium dodecanoate-d23, Potassium dodecanoate-D23, Potassium laurate-d23, Lauric-d23 acid potassium salt, Lauric acid-d23 potassium salt |
| InChI Formula | InChI=1 S/C12H24O2.K/c1-2-3-4-5-6-7-8-9-10-11-12(13)14;/h2-11H2,1H3,(H,13,14);/q;+1/p-1/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2; |
| SMILES | [K+].[2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)[O-] |
| IUPAC Name | potassium;2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosadeuteriododecanoate |
| Molecular Weight (g/mol) | 261.55 |
| Formula Weight | 261.278 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction