missing translation for 'onlineSavingsMsg'
Learn More

Potassium fluoride, Purum p.a., ≥99.0% (F), Honeywell™

Product Code. 16635873 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
10 kg
25 kg
Unit Size:
10kg
25kg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 16635873

Brand: Honeywell Research Chemicals 6024010KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-23-3
Molecular Formula FK
Molecular Weight (g/mol) 58.10
MDL Number MFCD00011398
InChI Key NROKBHXJSPEDAR-UHFFFAOYSA-M
IUPAC Name potassium fluoride
SMILES [F-].[K+]

Specifications

Boiling Point 1,505°C (1,013 hPa)
Melting Point 846°C
Color Colorless
CAS Max % 1
pH 8 to 9
Quantity 10 kg
Linear Formula KF
Beilstein 3902818
Grade Reagent
Density 2.48 g/cm3 (20°C)
Chemical Name or Material Potassium fluoride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.