missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hexacyanoferrate(Ii) Trihydrate, purum, Solstice

Product Code. 15681170
Change view
Click to view available options
Quantity:
25 kg
Unit Size:
25kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15681170 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15681170 Supplier Honeywell-Fluka Supplier No. 1263925KG
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

Purum,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Specifications

CAS 14459-95-1
Quantity 25 kg
Molecular Formula C6H6FeK4N6O3
Linear Formula K4Fe(CN)6 · 3H2O
MDL Number MFCD00167023
UN Number NONH for all modes of transport
InChI Key NLYKFUFIUWPPIZ-UHFFFAOYSA-N
SMILES O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N
IUPAC Name tetrapotassium hexacyanoironbis(ylium) trihydrate
Molecular Weight (g/mol) 422.39
Formula Weight 422.39g/mol
Percent Purity ≥99%
Grade purum
Chemical Name or Material Potassium Hexacyanoferrate(Ii) Trihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.