Learn More
Potassium hexacyanoferrate(III) solution, Honeywell™
Product Code. 15664140 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15664140 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15664140 Supplier Honeywell-Fluka Supplier No. 342721L
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Volumetric, 0.1 M K3Fe(CN)6
Honeywell´s high-quality titration products cover the complete titration workflow and include:
  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY
Specifications
CAS 13746-66-2, 7732-18-5
Quantity 1 L
MDL Number MFCD00011392
UN Number NONH for all modes of transport
Chemical Name or Material Potassium hexacyanoferrate(III) solution