missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hexacyanoferrate(III), TRC

Product Code. 30003934
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003934 10 g 10g
30003239 5 g 5g
30006623 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003934 Supplier Toronto Research Chemicals Supplier No. TRCP69508510G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13746-66-2
Molecular Formula C6FeK3N6
Molecular Weight (g/mol) 329.24
Synonym (OC-6-11)-Hexakis(cyano-kC)ferrate(3-) PotassiumHexacyanoferrate(3-) Ttripotassium (8CI), (OC-6-11)-Hexakis(cyano-C)ferrate(3-) Tripotassium, Hexakis(cyano-kC)Ferrate(3-) Tripotassium, Iron Potassium Cyanide (FeK3(CN)6), Potassium Cyanoferrate (K3Fe(CN)6), Potassium Ferricyanate, Potassium Ferricyanide, Potassium Ferricyanide (K3Fe(CN)6), Potassium Hexacyanoferrate (K3Fe(CN)6), Potassium Hexacyanoferrate(3-), Potassium Hexacyanoferrate(III), Potassium Iron Cyanide (K3Fe(CN)6), Red Prussiate, Tripotassium Ferric Hexacyanide, Tripotassium Ferricyanide, Tripotassium Hexacyanoferrate, Tripotassium Hexacyanoferrate(3-), Tripotassium Iron Hexacyanide
IUPAC Name tripotassium;hexakis(azanylidynemethyl)iron(3+)
SMILES [K+].[K+].[K+].N#[C-][Fe+3]([C-]#N)([C-]#N)([C-]#N)([C-]#N)[C-]#N

Specifications

Quantity 10 g
InChI Formula InChI=1S/6CN.Fe.3K/c6*1-2;;;;/q6*-1;+3;3*+1
Formula Weight 328.8445
Chemical Name or Material Potassium Hexacyanoferrate(III)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.