missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydrogen L-tartrate, 98+%

Product Code. 11478136
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11478136 5000 g 5000g
11468136 250 g 250g
11458136 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11478136 Supplier Thermo Scientific Alfa Aesar Supplier No. A15281.0I
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 868-14-4 | C4H5KO6 | 188.18 g/mol

Potassium hydrogen L-tartrate is used for stabilizing egg whites, whipped cream and for anti-caking and thickening. It is utilized for the prevention of crystallization of sugar syrups and reduces the discoloration of boiled vegetables. It is an active component in baking powder. It is used in household items in association with lemon juice or white vinegar to clean metals such as brass, aluminum and copper. Furthermore, it serves as a primary reference standard for a pH buffer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium hydrogen L-tartrate is used for stabilizing egg whites, whipped cream and for anti-caking and thickening. It is utilized for the prevention of crystallization of sugar syrups and reduces the discoloration of boiled vegetables. It is an active component in baking powder. It is used in household items in association with lemon juice or white vinegar to clean metals such as brass, aluminum and copper. Furthermore, it serves as a primary reference standard for a pH buffer.

Solubility
Soluble in water and dilute mineral acid. Insoluble in alcohol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 868-14-4
Molecular Formula C4H5KO6
Molecular Weight (g/mol) 188.18
MDL Number MFCD00065392,MFCD00071626,MFCD00064206
InChI Key KYKNRZGSIGMXFH-ZVGUSBNCSA-M
Synonym potassium bitartrate, l +-potassium hydrogen tartrate, potassium 3-carboxy-2,3-dihydroxypropanoate, monopotassium tartrate, potassium acid tartrate, kaliumtartrat, potassium hydrogentartrate, 1-potassium hydrogentartrate, ksc657e1t, butanedioic acid, 2,3-dihydroxy-2r,3r-, potassium salt 1:1
IUPAC Name 2,3-dihydroxybutanedioic acid;potassium
SMILES [K+].O[C@H]([C@@H](O)C([O-])=O)C(O)=O

Specifications

Melting Point ∼267°C (decomposition)
Density 1.954
Quantity 5000 g
Beilstein 6119985
Merck Index 14,7615
Solubility Information Soluble in water and dilute mineral acid. Insoluble in alcohol.
PubChem CID 376198361
Formula Weight 188.18
Percent Purity ≥98%
Chemical Name or Material Potassium hydrogen L-tartrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.